Assay Details
Gene Symbol: LOC4346142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792863.2 | XP_015648349.1 | 2 - 3 | 551 | 121 | |
GenBank mRNA | AK061214.1 | - | 2 - 3 | 496 | 121 | |
AK067802.1 | - | 2 - 3 | 497 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
