Assay Details
| Gene Symbol: LOC4346723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756508.2 | XP_015611994.1 | 1 - 2 | 492 | 58 | |
| GenBank mRNA | AK121426.1 | - | 1 - 2 | 313 | 58 | |
More Information
Set Membership: |
|

LOC4346723
Genomic Map