Assay Details
| Gene Symbol: LOC4346723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756508.2 | XP_015611994.1 | 7 - 8 | 1160 | 103 | |
| GenBank mRNA | AK121426.1 | - | 7 - 8 | 981 | 103 | |
More Information
Set Membership: |
|

LOC4346723
Genomic Map