Assay Details
| Gene Symbol: LOC4346723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756508.2 | XP_015611994.1 | 8 - 9 | 1262 | 87 | |
| GenBank mRNA | AK063871.1 | - | 2 - 3 | 136 | 87 | |
| AK121426.1 | - | 8 - 9 | 1083 | 87 | ||
More Information
Set Membership: |
|

LOC4346723
Genomic Map