Assay Details
| Gene Symbol: LOC4346937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755141.2 | XP_015610627.1 | 6 | 1507 | 149 | |
| GenBank mRNA | AK063210.1 | - | 1 | 91 | 149 | |
More Information
Set Membership: |
|

LOC4346937
Genomic Map