Assay Details
| Gene Symbol: LOC9271949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756627.1 | XP_015612113.1 | 1 - 2 | 143 | 94 | |
| GenBank mRNA | KT199371.1 | - | 1 - 2 | 104 | 94 | |
More Information
Set Membership: |
|

LOC9271949
Genomic Map