Assay Details
| Gene Symbol: LOC4347725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756948.2 | XP_015612434.1 | 2 - 3 | 2433 | 69 | |
| XM_015756949.2 | XP_015612435.1 | 2 - 3 | 2412 | 69 | ||
| GenBank mRNA | AK111927.1 | - | 2 - 3 | 452 | 69 | |
| AK112006.1 | - | 2 - 3 | 428 | 69 | ||
More Information
Set Membership: |
|

LOC4347725
Genomic Map