Assay Details
| Gene Symbol: LOC4347982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759562.2 | XP_015615048.1 | 1 | 3529 | 79 | |
| GenBank mRNA | AK243167.1 | - | 1 | 3536 | 79 | |
More Information
Set Membership: |
|

LOC4347982
Genomic Map