Assay Details
| Gene Symbol: LOC4348232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759477.2 | XP_015614963.1 | 1 - 2 | 180 | 109 | |
| GenBank mRNA | AK064839.1 | - | 1 - 2 | 158 | 109 | |
| AK066484.1 | - | 1 - 2 | 143 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015759477.2 transcripts.
Set Membership: |
|

LOC4348232
Genomic Map