Assay Details
| Gene Symbol: LOC4348232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759477.2 | XP_015614963.1 | 4 - 5 | 473 | 90 | |
| GenBank mRNA | AK064839.1 | - | 4 - 5 | 1532 | 90 | |
| AK066484.1 | - | 4 - 5 | 436 | 90 | ||
More Information
Set Membership: |
|

LOC4348232
Genomic Map