Assay Details
Gene Symbol: LOC4351561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762727.2 | XP_015618213.1 | 1 | 993 | 120 | |
GenBank mRNA | AK061582.1 | - | 1 | 840 | 120 | |
AK070481.1 | - | 1 | 951 | 120 | ||
AK104185.1 | - | 1 | 827 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
