Assay Details
| Gene Symbol: LOC4351781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763941.2 | XP_015619427.1 | 1 - 2 | 424 | 119 | |
| Gene Symbol: LOC9269803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763310.2 | XP_015618796.1 | 1 - 2 | 404 | 119 | |
More Information
Set Membership: |
|

LOC4351781
Genomic Map