Assay Details
| Gene Symbol: LOC9269121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764687.2 | XP_015620173.1 | 10 | 1751 | 83 | |
| GenBank mRNA | AK059945.1 | - | 5 | 910 | 83 | |
| AK060450.1 | - | 1 | 449 | 83 | ||
More Information
Set Membership: |
|

LOC9269121
Genomic Map