Assay Details
Gene Symbol: LOC4351869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763503.2 | XP_015618989.1 | 1 | 64 | 74 | |
GenBank mRNA | AK109396.2 | - | 1 | 48 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015763503.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
