Assay Details
| Gene Symbol: LOC4351887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763269.2 | XP_015618755.1 | 1 - 2 | 372 | 76 | |
| GenBank mRNA | AK066682.1 | - | 1 - 2 | 363 | 76 | |
| DQ243708.2 | - | 1 - 2 | 360 | 76 | ||
| DQ450957.1 | - | 1 - 2 | 36 | 76 | ||
More Information
Set Membership: |
|

LOC4351887
Genomic Map