Assay Details
| Gene Symbol: LOC4351887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763269.2 | XP_015618755.1 | 2 - 3 | 563 | 66 | |
| GenBank mRNA | DQ243708.2 | - | 2 - 3 | 551 | 66 | |
More Information
Set Membership: |
|

LOC4351887
Genomic Map