Assay Details
| Gene Symbol: LOC4352172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765036.2 | XP_015620522.1 | 1 - 2 | 509 | 92 | |
| GenBank mRNA | AK069528.1 | - | 1 - 2 | 468 | 92 | |
More Information
Set Membership: |
|

LOC4352172
Genomic Map