Assay Details
| Gene Symbol: LOC4352172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765036.2 | XP_015620522.1 | 2 - 3 | 606 | 99 | |
| GenBank mRNA | AK069528.1 | - | 2 - 3 | 565 | 99 | |
More Information
Set Membership: |
|

LOC4352172
Genomic Map