Assay Details
| Gene Symbol: LOC4352407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763297.2 | XP_015618783.1 | 1 - 2 | 211 | 107 | |
| GenBank mRNA | AK241686.1 | - | 1 - 2 | 210 | 107 | |
More Information
Set Membership: |
|

LOC4352407
Genomic Map