Assay Details
| Gene Symbol: LOC4352808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764691.2 | XP_015620177.1 | 20 - 21 | 3327 | 67 | |
| GenBank mRNA | AK062131.1 | - | 4 - 5 | 383 | 67 | |
| AK065064.1 | - | 20 - 21 | 3024 | 67 | ||
| AK066418.1 | - | 20 - 21 | 3297 | 67 | ||
More Information
Set Membership: |
|

LOC4352808
Genomic Map