Assay Details
| Gene Symbol: LOC4352808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764691.2 | XP_015620177.1 | 5 - 6 | 1127 | 70 | |
| GenBank mRNA | AK065064.1 | - | 5 - 6 | 824 | 70 | |
| AK066418.1 | - | 5 - 6 | 1097 | 70 | ||
More Information
Set Membership: |
|

LOC4352808
Genomic Map