Assay Details
| Gene Symbol: LOC4352821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763589.2 | XP_015619075.1 | 6 - 7 | 2153 | 88 | |
| GenBank mRNA | AK111785.1 | - | 6 - 7 | 2154 | 88 | |
More Information
Set Membership: |
|

LOC4352821
Genomic Map