Assay Details
| Gene Symbol: LOC4352873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763005.2 | XP_015618491.1 | 1 - 2 | 2749 | 67 | |
| GenBank mRNA | AK071957.1 | - | 1 - 2 | 2744 | 67 | |
More Information
Set Membership: |
|

LOC4352873
Genomic Map