Assay Details
Gene Symbol: LOC4350166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759693.2 | XP_015615179.1 | 2 - 3 | 384 | 125 | |
GenBank mRNA | AK099694.1 | - | 2 - 3 | 369 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
