Assay Details
| Gene Symbol: LOC9266255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760936.2 | XP_015616422.1 | 1 - 2 | 164 | 67 | |
| XM_015760938.2 | XP_015616424.1 | 1 - 2 | 167 | 67 | ||
| GenBank mRNA | AK241350.1 | - | 1 - 2 | 83 | 67 | |
| Gene Symbol: LOC4350701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760927.2 | XP_015616413.1 | 2 - 3 | 541 | 67 | |
| XM_015760928.2 | XP_015616414.1 | 2 - 3 | 523 | 67 | ||
| GenBank mRNA | AK060533.1 | - | 1 - 2 | 163 | 67 | |
More Information
Set Membership: |
|

LOC9266255
Genomic Map