Assay Details
| Gene Symbol: LOC4350701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760927.2 | XP_015616413.1 | 6 - 7 | 1024 | 91 | |
| XM_015760928.2 | XP_015616414.1 | 7 - 8 | 913 | 91 | ||
| GenBank mRNA | AK060533.1 | - | 6 - 7 | 553 | 91 | |
More Information
Set Membership: |
|

LOC4350701
Genomic Map