Assay Details
| Gene Symbol: LOC4351051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761745.2 | XP_015617231.1 | 2 - 3 | 287 | 68 | |
| GenBank mRNA | AK059959.1 | - | 2 - 3 | 262 | 68 | |
More Information
Set Membership: |
|

LOC4351051
Genomic Map