Assay Details
| Gene Symbol: LOC4351130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760546.2 | XP_015616032.1 | 1 - 2 | 1297 | 61 | |
| GenBank mRNA | AK070275.1 | - | 1 - 2 | 1292 | 61 | |
More Information
Set Membership: |
|

LOC4351130
Genomic Map