Assay Details
| Gene Symbol: LOC4326151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759924.2 | XP_015615410.1 | 29 - 30 | 3635 | 68 | |
| XM_015759931.2 | XP_015615417.1 | 29 - 30 | 3635 | 68 | ||
| XM_015759946.2 | XP_015615432.1 | 15 - 16 | 1981 | 68 | ||
More Information
Set Membership: |
|

LOC4326151
Genomic Map