Assay Details
| Gene Symbol: LOC4326151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759924.2 | XP_015615410.1 | 45 - 46 | 5605 | 114 | |
| XM_015759931.2 | XP_015615417.1 | 45 - 46 | 5605 | 114 | ||
| XM_015759946.2 | XP_015615432.1 | 31 - 32 | 3951 | 114 | ||
More Information
Set Membership: |
|

LOC4326151
Genomic Map