Assay Details
| Gene Symbol: LOC4325197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792510.1 | XP_015647996.1 | 12 - 13 | 2391 | 88 | |
| GenBank mRNA | AK120051.1 | - | 12 - 13 | 2393 | 88 | |
| AK121259.1 | - | 1 - 2 | 90 | 88 | ||
More Information
Set Membership: |
|

LOC4325197
Genomic Map