Assay Details
| Gene Symbol: LOC4325197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792510.1 | XP_015647996.1 | 13 - 14 | 2530 | 113 | |
| GenBank mRNA | AK120051.1 | - | 13 - 14 | 2532 | 113 | |
| AK121259.1 | - | 2 - 3 | 229 | 113 | ||
More Information
Set Membership: |
|

LOC4325197
Genomic Map