Assay Details
| Gene Symbol: LOC4325197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792510.1 | XP_015647996.1 | 18 - 19 | 3178 | 81 | |
| GenBank mRNA | AK120051.1 | - | 18 - 19 | 3180 | 81 | |
| AK121259.1 | - | 7 - 8 | 877 | 81 | ||
More Information
Set Membership: |
|

LOC4325197
Genomic Map