Assay Details
| Gene Symbol: LOC4325197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792510.1 | XP_015647996.1 | 3 - 4 | 1143 | 87 | |
| GenBank mRNA | AK120051.1 | - | 3 - 4 | 1145 | 87 | |
More Information
Set Membership: |
|

LOC4325197
Genomic Map