Assay Details
| Gene Symbol: LOC4327119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791158.2 | XP_015646644.1 | 2 - 3 | 417 | 56 | |
| GenBank mRNA | AK287596.1 | - | 2837 | 56 | ||
| AK287669.1 | - | 2 - 3 | 352 | 56 | ||
More Information
Set Membership: |
|

LOC4327119
Genomic Map