Assay Details
| Gene Symbol: LOC4325344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758092.2 | XP_015613578.1 | 8 - 9 | 1493 | 139 | |
| XM_015758100.2 | XP_015613586.1 | 8 - 9 | 1493 | 139 | ||
| GenBank mRNA | AK060699.1 | - | 8 - 9 | 1458 | 139 | |
More Information
Set Membership: |
|

LOC4325344
Genomic Map