Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 12 - 13 | 1399 | 70 | |
| GenBank mRNA | AK060358.1 | - | 5 - 6 | 342 | 70 | |
| AK072842.1 | - | 12 - 13 | 1307 | 70 | ||
| AK121770.1 | - | 12 - 13 | 1309 | 70 | ||
| KC140122.1 | - | 12 - 13 | 1274 | 70 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map