Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 15 - 16 | 1642 | 126 | |
| GenBank mRNA | AK060358.1 | - | 8 - 9 | 585 | 126 | |
| AK072842.1 | - | 15 - 16 | 1550 | 126 | ||
| AK121770.1 | - | 15 - 16 | 1552 | 126 | ||
| KC140122.1 | - | 15 - 16 | 1517 | 126 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map