Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 18 - 19 | 1884 | 76 | |
| GenBank mRNA | AK060358.1 | - | 11 - 12 | 827 | 76 | |
| AK072842.1 | - | 18 - 19 | 1792 | 76 | ||
| AK121770.1 | - | 18 - 19 | 1794 | 76 | ||
| KC140122.1 | - | 18 - 19 | 1759 | 76 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map