Assay Details
| Gene Symbol: LOC4326769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766067.2 | XP_015621553.1 | 9 - 10 | 1171 | 80 | |
| GenBank mRNA | AK060358.1 | - | 2 - 3 | 114 | 80 | |
| AK072842.1 | - | 9 - 10 | 1081 | 80 | ||
| AK121770.1 | - | 9 - 10 | 1081 | 80 | ||
| KC140122.1 | - | 9 - 10 | 1046 | 80 | ||
More Information
Set Membership: |
|

LOC4326769
Genomic Map