Assay Details
| Gene Symbol: LOC4326774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768209.2 | XP_015623695.1 | 1 - 2 | 485 | 69 | |
| GenBank mRNA | AK288019.1 | - | 1 - 2 | 411 | 69 | |
More Information
Set Membership: |
|

LOC4326774
Genomic Map