Assay Details
| Gene Symbol: LOC4327329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755947.2 | XP_015611433.1 | 7 - 8 | 1867 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327329| Gene Symbol: LOC4327329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755947.2 | XP_015611433.1 | 7 - 8 | 1867 | 96 | |
Set Membership: |
|