Assay Details
| Gene Symbol: LOC4327180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793852.1 | XP_015649338.1 | 3 - 4 | 367 | 65 | |
| GenBank mRNA | AK059958.1 | - | 3 - 4 | 285 | 65 | |
| AK070320.1 | - | 3 - 4 | 297 | 65 | ||
More Information
Set Membership: |
|

LOC4327180
Genomic Map