Assay Details
| Gene Symbol: LOC4324288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775240.2 | XP_015630726.1 | 6 - 7 | 1177 | 115 | |
| GenBank mRNA | AK063476.1 | - | 6 - 7 | 1168 | 115 | |
More Information
Set Membership: |
|

LOC4324288
Genomic Map