Assay Details
| Gene Symbol: LOC9268236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757546.2 | XP_015613032.1 | 3 - 4 | 982 | 120 | |
| GenBank mRNA | AK287868.1 | - | 3 - 4 | 968 | 120 | |
More Information
Set Membership: |
|

LOC9268236
Genomic Map