Assay Details
| Gene Symbol: LOC4327702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765578.2 | XP_015621064.1 | 13 - 14 | 1744 | 80 | |
| XM_015765580.2 | XP_015621066.1 | 13 - 14 | 1744 | 80 | ||
| GenBank mRNA | AK064212.1 | - | 1 - 2 | 152 | 80 | |
More Information
Set Membership: |
|

LOC4327702
Genomic Map