Assay Details
| Gene Symbol: LOC4327702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765578.2 | XP_015621064.1 | 14 - 15 | 2004 | 69 | |
| XM_015765580.2 | XP_015621066.1 | 14 - 15 | 2004 | 69 | ||
| GenBank mRNA | AK064212.1 | - | 2 - 3 | 412 | 69 | |
More Information
Set Membership: |
|

LOC4327702
Genomic Map