Assay Details
| Gene Symbol: LOC4326755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765686.1 | XP_015621172.1 | 1 - 2 | 211 | 96 | |
| GenBank mRNA | AK120732.1 | - | 1 - 2 | 122 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765686.1 transcripts.
Set Membership: |
|

LOC4326755
Genomic Map