Assay Details
| Gene Symbol: LOC4326204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755790.2 | XP_015611276.1 | 4 - 5 | 710 | 122 | |
| GenBank mRNA | AK071712.1 | - | 4 - 5 | 715 | 122 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC4326204
Genomic Map