Assay Details
| Gene Symbol: LOC4326812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766814.2 | XP_015622300.1 | 13 - 14 | 2379 | 99 | |
| XM_015766815.2 | XP_015622301.1 | 12 - 13 | 3332 | 99 | ||
| GenBank mRNA | AY332479.1 | - | 13 - 14 | 2208 | 99 | |
More Information
Set Membership: |
|

LOC4326812
Genomic Map